accessibility menu, dialog, popup

UserName

CAS RN 1528-74-1

ONONOO

CAS RN 1528-74-1

IUPAC Name: 4,4'-dinitro-1,1'-biphenyl
Synonyms: 4,4'-dinitrobiphenyl
Molecular Weight (g/mol): 244.21
Molecular Formula: C12H8N2O4
InChi Key: BDLNCFCZHNKBGI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results