missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzophenone, 98+%

Catalog No. p-4187322
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408220010 1 g Glass bottle
AC408220100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC408220010 Supplier Thermo Scientific Chemicals Supplier No. 408220010
Only null left

CAS: 611-97-2 | C15H14O | 210.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-97-2
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00017214
InChI Key ZWPWLKXZYNXATK-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzophenone, p-tolyl ketone, bis 4-methylphenyl methanone, di-p-tolyl ketone, methanone, bis 4-methylphenyl, p,p'-dimethylbenzophenone, p,p'-dimethyl di-phenyl ketone, bis-p-tolylmethanone, benzophenone, 4,4'-dimethyl, 4,4-dimethylbenzophenone
PubChem CID 69148
IUPAC Name bis(4-methylphenyl)methanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 93°C to 97°C
Color Brown
Boiling Point 200°C (17.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 210.28
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethylbenzophenone
Show More Show Less
EINECSNumber 210-287-6

RUO – Research Use Only