accessibility menu, dialog, popup

UserName

CH3OH3C

CAS RN 611-97-2

IUPAC Name: bis(4-methylphenyl)methanone
Synonyms: bis(4-methylphenyl)methanone
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: ZWPWLKXZYNXATK-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)C1=CC=C(C)C=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)