missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzhydrol 98.0+%, TCI America™

Catalog No. D54601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54601G 1 g
D54605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54601G Supplier TCI America Supplier No. D54601G

Chemical Identifiers

CAS 885-77-8
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.292
MDL Number MFCD00017216
InChI Key RGYZQSCFKFDECZ-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzhydrol, di-p-tolylmethanol, bis 4-methylphenyl methanol, benzenemethanol, 4-methyl-.alpha.-4-methylphenyl, benzhydrol, 4,4'-dimethyl, di p-tolyl methanol, 4,4-dimethylbenzhydrol, 4,4'-ditolyl carbinol, benzhydrol,4'-dimethyl
PubChem CID 279356
IUPAC Name bis(4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)O

Specifications

Melting Point 71°C
Color White
Quantity 1 g
Formula Weight 212.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethylbenzhydrol