accessibility menu, dialog, popup

UserName

CAS RN 885-77-8

IUPAC Name: bis(4-methylphenyl)methanol
Synonyms: bis(4-methylphenyl)methanol
Molecular Weight (g/mol): 212.29
Molecular Formula: C15H16O
InChi Key: RGYZQSCFKFDECZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)