Learn More
4,4'-Dimethylbenzhydrol, 98%
CAS: 885-77-8 | C15H16O | 212.292 g/mol
Supplier: Thermo Scientific Chemicals L0590406
Description
4,4'-Dimethylbenzhydrol is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,4′-Dimethylbenzhydrol is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
| 4,4'-Dimethylbenzhydrol | |
| 69°C to 73°C | |
| MFCD00017216 | |
| 1874128 | |
| Insoluble in water. | |
| CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)O | |
| 212.292 | |
| 212.29 |
| 885-77-8 | |
| C15H16O | |
| 5 g | |
| 4,4'-dimethylbenzhydrol, di-p-tolylmethanol, bis 4-methylphenyl methanol, benzenemethanol, 4-methyl-.alpha.-4-methylphenyl, benzhydrol, 4,4'-dimethyl, di p-tolyl methanol, 4,4-dimethylbenzhydrol, 4,4'-ditolyl carbinol, benzhydrol,4'-dimethyl | |
| RGYZQSCFKFDECZ-UHFFFAOYSA-N | |
| bis(4-methylphenyl)methanol | |
| 279356 | |
| 98% |
Chemical Identifiers
| 885-77-8 | |
| 212.292 | |
| RGYZQSCFKFDECZ-UHFFFAOYSA-N | |
| 279356 | |
| CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)O |
| C15H16O | |
| MFCD00017216 | |
| 4,4'-dimethylbenzhydrol, di-p-tolylmethanol, bis 4-methylphenyl methanol, benzenemethanol, 4-methyl-.alpha.-4-methylphenyl, benzhydrol, 4,4'-dimethyl, di p-tolyl methanol, 4,4-dimethylbenzhydrol, 4,4'-ditolyl carbinol, benzhydrol,4'-dimethyl | |
| bis(4-methylphenyl)methanol |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only