missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethylbenzhydrol, 98%

Catalog No. AAL0590403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0590403 1 g
AAL0590406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0590403 Supplier Thermo Scientific Chemicals Supplier No. L0590403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 885-77-8 | C15H16O | 212.292 g/mol

4,4'-Dimethylbenzhydrol is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethylbenzhydrol is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 885-77-8
Molecular Formula C15H16O
Molecular Weight (g/mol) 212.292
MDL Number MFCD00017216
InChI Key RGYZQSCFKFDECZ-UHFFFAOYSA-N
Synonym 4,4'-dimethylbenzhydrol, di-p-tolylmethanol, bis 4-methylphenyl methanol, benzenemethanol, 4-methyl-.alpha.-4-methylphenyl, benzhydrol, 4,4'-dimethyl, di p-tolyl methanol, 4,4-dimethylbenzhydrol, 4,4'-ditolyl carbinol, benzhydrol,4'-dimethyl
PubChem CID 279356
IUPAC Name bis(4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)C(C2=CC=C(C=C2)C)O

Specifications

Melting Point 69°C to 73°C
Quantity 1 g
Beilstein 1874128
Solubility Information Insoluble in water.
Formula Weight 212.29
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethylbenzhydrol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only