missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxydiphenylamine 98.0+%, TCI America™

Catalog No. p-7126333
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06321G 1 g
D06325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06321G Supplier TCI America Supplier No. D06321G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-70-2
Molecular Formula C14H15NO2
Molecular Weight (g/mol) 229.28
MDL Number MFCD00014895
InChI Key VCOONNWIINSFBA-UHFFFAOYSA-N
Synonym bis 4-methoxyphenyl amine, 4,4'-dimethoxydiphenylamine, di-p-anisylamine, di-p-methoxyphenylamine, 4-methoxy-n-4-methoxyphenyl aniline, bis p-anisyl amine, bis p-methoxyphenyl amine, benzenamine, 4-methoxy-n-4-methoxyphenyl, termofleks a, bis-4-methoxy-phenyl-amine
PubChem CID 7571
IUPAC Name 4-methoxy-N-(4-methoxyphenyl)aniline
SMILES COC1=CC=C(NC2=CC=C(OC)C=C2)C=C1

Specifications

Melting Point 103°C
Color Yellow
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxydiphenylamine
EINECSNumber (3)-2783
RTECSNumber DU9085000
TSCA Yes