accessibility menu, dialog, popup

UserName

H3CONHOCH3

CAS RN 101-70-2

IUPAC Name: 4-methoxy-N-(4-methoxyphenyl)aniline
Synonyms: 4-methoxy-N-(4-methoxyphenyl)aniline
Molecular Weight (g/mol): 229.28
Molecular Formula: C14H15NO2
InChi Key: VCOONNWIINSFBA-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC2=CC=C(OC)C=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)