missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxydiphenylamine, 98%

Catalog No. AAL0132203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0132203 1 g
AAL0132206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0132203 Supplier Thermo Scientific Chemicals Supplier No. L0132203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 101-70-2 | C14H15NO2 | 229.28 g/mol

4,4'-Dimethoxydiphenylamine is a diphenylamine derivative used as a chemical additive for cured rubber. It is used in the synthesis of amine electron donating groups in organic donor-bridge-acceptor dyads in Dye Sensitized Solar Cells (DSSC). 4,4?-Dimethoxydiphenylamine is an electron donor and used in the synthesis of organic nonlinear optical (NLO) chromophores which exhibit high thermal and chemical stability, high degree of non-linearity and transparency. They are used in high performance photonic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxydiphenylamine is a diphenylamine derivative used as a chemical additive for cured rubber. It is used in the synthesis of amine electron donating groups in organic donor-bridge-acceptor dyads in Dye Sensitized Solar Cells (DSSC). 4,4′-Dimethoxydiphenylamine is an electron donor and used in the synthesis of organic nonlinear optical (NLO) chromophores which exhibit high thermal and chemical stability, high degree of non-linearity and transparency. They are used in high performance photonic devices.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Air Sensitive. Store under dry inert gas. Incompatible with isocyanates, halogenated organics, peroxides, phenols (acidic), epoxides, anhydrides, and acid halides.

Chemical Identifiers

CAS 101-70-2
Molecular Formula C14H15NO2
Molecular Weight (g/mol) 229.28
MDL Number MFCD00014895
InChI Key VCOONNWIINSFBA-UHFFFAOYSA-N
Synonym bis 4-methoxyphenyl amine, 4,4'-dimethoxydiphenylamine, di-p-anisylamine, di-p-methoxyphenylamine, 4-methoxy-n-4-methoxyphenyl aniline, bis p-anisyl amine, bis p-methoxyphenyl amine, benzenamine, 4-methoxy-n-4-methoxyphenyl, termofleks a, bis-4-methoxy-phenyl-amine
PubChem CID 7571
IUPAC Name 4-methoxy-N-(4-methoxyphenyl)aniline
SMILES COC1=CC=C(NC2=CC=C(OC)C=C2)C=C1

Specifications

Melting Point 100°C to 104°C
Quantity 1 g
Beilstein 2214262
Sensitivity Air Sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 229.28
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethoxydiphenylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-968-1
RTECSNumber DU9085000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only