missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzhydrylamine 98.0+%, TCI America™

Catalog No. D22025G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22025G 5 g
D22021G 1 g
D220225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D22025G Supplier TCI America Supplier No. D22025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19293-62-0
Molecular Formula C15H17NO2
Molecular Weight (g/mol) 243.31
MDL Number MFCD00191409
InChI Key HROGQYMZWGPHIB-UHFFFAOYSA-N
Synonym 1,1-Di(p-anisyl)methylamine
PubChem CID 350042
IUPAC Name 1,1-bis(4-methoxyphenyl)methanamine
SMILES COC1=CC=C(C=C1)C(N)C1=CC=C(OC)C=C1

Specifications

Melting Point 62°C
Color White
Quantity 5 g
Formula Weight 243.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxybenzhydrylamine