accessibility menu, dialog, popup

UserName

CAS RN 19293-62-0

H3CONH2OCH3

CAS RN 19293-62-0

IUPAC Name: 1,1-bis(4-methoxyphenyl)methanamine
Synonyms: 1,1-bis(4-methoxyphenyl)methanamine
Molecular Weight (g/mol): 243.31
Molecular Formula: C15H17NO2
InChi Key: HROGQYMZWGPHIB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(N)C1=CC=C(OC)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results