missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxybenzhydrol, 98+%

Catalog No. AAA1460409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1460409 10 g
AAA1460418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1460409 Supplier Thermo Scientific Chemicals Supplier No. A1460409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 728-87-0 | C15H16O3 | 244.29 g/mol

4,4?-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethoxybenzhydrol was used to study the kinetics and mechanism of oxidation of substituted benzhydrols to corresponding benzophenones in the presence of Hg(OAc)2 by N-bromosuccinimide. It is useful in the protection of glutamine and asparagine residues in peptide synthesis, reagent for the N-protection of amides.

Solubility
Soluble in water (0.44 g/L @ 25°C), methanol (0.1 g/mL).

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 728-87-0
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00008410
InChI Key ZODAOVNETBTTJX-UHFFFAOYSA-N
Synonym 4,4'-dimethoxybenzhydrol, bis 4-methoxyphenyl methanol, p,p'-dimethoxybenzhydryl alcohol, p,p'-dimethoxybenzhydrol, bis p-methoxyphenyl carbinol, benzenemethanol, 4-methoxy-.alpha.-4-methoxyphenyl, 4-methoxy-4'-methoxybenzhydrol, bis 4-methoxyphenyl methan-1-ol, benzhydrol, 4,4'-dimethoxy, acmc-209ooh
PubChem CID 69768
IUPAC Name bis(4-methoxyphenyl)methanol
SMILES COC1=CC=C(C=C1)C(O)C1=CC=C(OC)C=C1

Specifications

Melting Point 69°C to 71°C
Quantity 10 g
Beilstein 748532
Solubility Information Soluble in water (0.44g/L @ 25°C),methanol (0.1g/mL).
Formula Weight 244.29
Percent Purity ≥98%
Chemical Name or Material 4,4'-Dimethoxybenzhydrol
EINECSNumber 211-975-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only