missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethoxy-trans-stilbene 98.0+%, TCI America™

Catalog No. p-7127606
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3104500MG 500 mg
D31041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3104500MG Supplier TCI America Supplier No. D3104500MG

Chemical Identifiers

CAS 15638-14-9
Molecular Formula C16H16O2
Molecular Weight (g/mol) 240.302
InChI Key CAWFCZIEFIQKRV-ONEGZZNKSA-N
Synonym bianisal, 4,4'-dimethoxystilbene, bianisylidene, photoanethole, p,p'-dimethoxystilbene, 4,4'-dimethoxy-trans-stilbene, e-1,2-bis 4-methoxyphenyl ethene, 1-methoxy-4-e-2-4-methoxyphenyl ethenyl benzene, 4,4/'-dimethoxystilbene, 1,1'-e-ethene-1,2-diylbis 4-methoxybenzene
PubChem CID 641296
IUPAC Name 1-methoxy-4-[(E)-2-(4-methoxyphenyl)ethenyl]benzene
SMILES COC1=CC=C(C=C1)C=CC2=CC=C(C=C2)OC

Specifications

Melting Point 214°C
Quantity 500 mg
Formula Weight 240.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxy-trans-stilbene
TSCA No
Recommended Storage Refrigerator