accessibility menu, dialog, popup

UserName

H3COOCH3(E)

CAS RN 15638-14-9

IUPAC Name: 1-methoxy-4-[(1E)-2-(4-methoxyphenyl)ethenyl]benzene
Synonyms: 1-methoxy-4-[(1E)-2-(4-methoxyphenyl)ethenyl]benzene
Molecular Weight (g/mol): 240.3
Molecular Formula: C16H16O2
InChi Key: CAWFCZIEFIQKRV-ONEGZZNKSA-N
SMILES: COC1=CC=C(\C=C\C2=CC=C(OC)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results