missing translation for 'onlineSavingsMsg'
Learn More

4,4″-Diiodo-p-terphenyl 98.0+%, TCI America™

$253.51 - $754.94

Chemical Identifiers

CAS 19053-14-6
Molecular Formula C18H12I2
Molecular Weight (g/mol) 482.103
InChI Key QGMMWGLDOBFHTL-UHFFFAOYSA-N
PubChem CID 3563583
IUPAC Name 1,4-bis(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)C3=CC=C(C=C3)I
View More Specs

Products 2
Catalog Number Mfr. No. missing translation for 'Quantity' Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D35341G
View Documents
TCI America
D35341G
1 g
Each for $253.51
Only null left
 
D35345G
View Documents
TCI America
D35345G
5 g
Each for $754.94
Only null left
 
Description

Description

Specifications

Chemical Identifiers

19053-14-6
482.103
3563583
C1=CC(=CC=C1C2=CC=C(C=C2)I)C3=CC=C(C=C3)I
C18H12I2
QGMMWGLDOBFHTL-UHFFFAOYSA-N
1,4-bis(4-iodophenyl)benzene

Specifications

19053-14-6
C18H12I2
3152
C1=CC(=CC=C1C2=CC=C(C=C2)I)C3=CC=C(C=C3)I
482.103
482.10
Crystalline Powder
328°C
1 g
QGMMWGLDOBFHTL-UHFFFAOYSA-N
1,4-bis(4-iodophenyl)benzene
3563583
≥98.0% (GC)
4,4′′-Diiodo-p-terphenyl
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'