accessibility menu, dialog, popup

UserName

II

CAS RN 19053-14-6

IUPAC Name: 4-iodo-4'-(4-iodophenyl)-1,1'-biphenyl
Synonyms: 4-iodo-4'-(4-iodophenyl)-1,1'-biphenyl
Molecular Weight (g/mol): 482.1
Molecular Formula: C18H12I2
InChi Key: QGMMWGLDOBFHTL-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results