missing translation for 'onlineSavingsMsg'
Learn More

4,4″-Diiodo-p-terphenyl 98.0+%, TCI America™

Catalog No. D35345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35345G 5 g
D35341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35345G Supplier TCI America Supplier No. D35345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19053-14-6
Molecular Formula C18H12I2
Molecular Weight (g/mol) 482.103
InChI Key QGMMWGLDOBFHTL-UHFFFAOYSA-N
PubChem CID 3563583
IUPAC Name 1,4-bis(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)C3=CC=C(C=C3)I

Specifications

Melting Point 328°C
Quantity 5 g
UN Number 3152
Formula Weight 482.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′′-Diiodo-p-terphenyl