missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diiodo-3,3'-dimethylbiphenyl 98.0+%, TCI America™
Supplier: TCI America D27595G
Specifications
| 4,4′-Diiodo-3,3′-dimethylbiphenyl | |
| 108°C | |
| C14H12I2 | |
| 5 g | |
| CC1=C(C=CC(=C1)C2=CC(=C(C=C2)I)C)I | |
| 434.059 | |
| 434.06 | |
| Crystalline Powder |
| 7583-27-9 | |
| Yellow | |
| MFCD04038414 | |
| ISWXEMYZGWXIIZ-UHFFFAOYSA-N | |
| 1-iodo-4-(4-iodo-3-methylphenyl)-2-methylbenzene | |
| 22323609 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 7583-27-9 | |
| 434.059 | |
| ISWXEMYZGWXIIZ-UHFFFAOYSA-N | |
| 1-iodo-4-(4-iodo-3-methylphenyl)-2-methylbenzene |
| C14H12I2 | |
| MFCD04038414 | |
| 22323609 | |
| CC1=C(C=CC(=C1)C2=CC(=C(C=C2)I)C)I |
Safety and Handling
TSCA : No