missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diiodo-3,3'-dimethylbiphenyl 98.0+%, TCI America™

Catalog No. p-7127480
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27595G 5 g
D275925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27595G Supplier TCI America Supplier No. D27595G

Chemical Identifiers

CAS 7583-27-9
Molecular Formula C14H12I2
Molecular Weight (g/mol) 434.059
MDL Number MFCD04038414
InChI Key ISWXEMYZGWXIIZ-UHFFFAOYSA-N
PubChem CID 22323609
IUPAC Name 1-iodo-4-(4-iodo-3-methylphenyl)-2-methylbenzene
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)I)C)I

Specifications

Melting Point 108°C
Color Yellow
Quantity 5 g
Formula Weight 434.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diiodo-3,3′-dimethylbiphenyl