missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxydiphenylmethane 99.0+%, TCI America™

Catalog No. p-7118688
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08195G 5 g
B081925G 25 g
B0819100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B08195G Supplier TCI America Supplier No. B08195G
Only null left

Chemical Identifiers

CAS 620-92-8
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00002385
InChI Key PXKLMJQFEQBVLD-UHFFFAOYSA-N
Synonym 4,4'-methylenediphenol, bis 4-hydroxyphenyl methane, 4,4'-dihydroxydiphenylmethane, phenol, 4,4'-methylenebis, p-p-hydroxybenzyl phenol, bis p-hydroxyphenyl methane, phenol, 4,4'-methylenedi, 4,4'-methylenebisphenol, p,p'-bis hydroxyphenyl methane, 4-4-hydroxyphenyl methyl phenol
PubChem CID 12111
ChEBI CHEBI:34575
IUPAC Name 4-[(4-hydroxyphenyl)methyl]phenol
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)O)O

Specifications

Melting Point 160°C
Quantity 5 g
Formula Weight 200.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxydiphenylmethane
EINECSNumber (4)-0090
RTECSNumber SL9625000
TSCA Yes