missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxydiphenyl Ether 98.0+%, TCI America™

Catalog No. D2121500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2121500G 500 g
D212125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2121500G Supplier TCI America Supplier No. D2121500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1965-09-9
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.21
MDL Number MFCD00016463
InChI Key NZGQHKSLKRFZFL-UHFFFAOYSA-N
Synonym 4,4'-oxydiphenol, 4,4'-dihydroxydiphenyl ether, phenol, 4,4'-oxybis, 4-4-hydroxyphenoxy phenol, p,p'-oxydiphenol, phenol, 4,4'-oxydi, p,p'-oxybisphenol, 4,4'-oxybisphenol, bis 4-hydroxyphenyl ether, unii-7l83kgh26s
PubChem CID 16069
IUPAC Name 4-(4-hydroxyphenoxy)phenol
SMILES C1=CC(=CC=C1O)OC2=CC=C(C=C2)O

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 500 g
Formula Weight 202.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxydiphenyl Ether
EINECSNumber (3)-0654
RTECSNumber SM6040000
TSCA Yes