accessibility menu, dialog, popup

UserName

CAS RN 1965-09-9

IUPAC Name: 4-(4-hydroxyphenoxy)phenol
Synonyms: phenol, 4,4'-oxybis-
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: NZGQHKSLKRFZFL-UHFFFAOYSA-N
SMILES: OC1=CC=C(OC2=CC=C(O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results