missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxybenzophenone 98.0+%, TCI America™

Catalog No. p-7126298
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D057725G 25 g
D0577100G 100 g
D0577500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D057725G Supplier TCI America Supplier No. D057725G
Only null left

Chemical Identifiers

CAS 611-99-4
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002358
InChI Key RXNYJUSEXLAVNQ-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybenzophenone, bis 4-hydroxyphenyl methanone, hbp ketone, methanone, bis 4-hydroxyphenyl, bis 4-hydroxyphenyl ketone, 4,4'-dihydoxy-benzophenone, bis p-hydroxy benzophenone, p,p'-dihydroxybenzophenone, 4,4'-dihydroxydiphenyl ketone, benzophenone, 4,4'-dihydroxy
PubChem CID 69150
ChEBI CHEBI:34365
IUPAC Name bis(4-hydroxyphenyl)methanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)O)O

Specifications

Melting Point 221°C
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxybenzophenone
EINECSNumber (4)-0132
RTECSNumber DJ0880000
TSCA No