missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dihydroxyazobenzene 98.0+%, TCI America™

Catalog No. D34315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34315G 5 g
D34311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34315G Supplier TCI America Supplier No. D34315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2050-16-0
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.22
MDL Number MFCD00045778
InChI Key LJIUQBUUZNTUQK-UHFFFAOYSA-N
PubChem CID 5385293
IUPAC Name 4-[2-(4-hydroxyphenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC=C(NN=C2C=CC(=O)C=C2)C=C1

Specifications

Melting Point 215°C
Quantity 5 g
Synonym 4,4′-Azodiphenol
Formula Weight 214.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dihydroxyazobenzene