accessibility menu, dialog, popup

UserName

CAS RN 2050-16-0

IUPAC Name: 4-[2-(4-hydroxyphenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: 4-[2-(4-hydroxyphenyl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: LJIUQBUUZNTUQK-UHFFFAOYSA-N
SMILES: OC1=CC=C(NN=C2C=CC(=O)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results