missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzhydryl Chloride 97.0+%, TCI America™

Catalog No. p-7128120
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41385G 5 g
D413825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41385G Supplier TCI America Supplier No. D41385G
Only null left

Chemical Identifiers

CAS 27064-94-4
Molecular Formula C13H9ClF2
Molecular Weight (g/mol) 238.662
MDL Number MFCD00044329
InChI Key FHPNLCLHMNPLEW-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
PubChem CID 2723773
IUPAC Name 1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
UN Number 3265
Formula Weight 238.66
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4,4′-Difluorobenzhydryl Chloride