accessibility menu, dialog, popup

UserName

FClF

CAS RN 27064-94-4

IUPAC Name: 1-[chloro(4-fluorophenyl)methyl]-4-fluorobenzene
Synonyms: 1-[chloro(4-fluorophenyl)methyl]-4-fluorobenzene
Molecular Weight (g/mol): 238.66
Molecular Formula: C13H9ClF2
InChi Key: FHPNLCLHMNPLEW-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(Cl)C1=CC=C(F)C=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)