missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Difluorobenzhydryl chloride, 98%

Catalog No. AAA1122606
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1122606 5 g
AAA1122609 10 g
AAA1122618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1122606 Supplier Thermo Scientific Chemicals Supplier No. A1122606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27064-94-4 | C13H9ClF2 | 238.662 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27064-94-4
Molecular Formula C13H9ClF2
Molecular Weight (g/mol) 238.662
MDL Number MFCD00044329
InChI Key FHPNLCLHMNPLEW-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzhydryl chloride, 4,4'-chloromethylene bis fluorobenzene, 1,1'-chloromethylene bis 4-fluorobenzene, chlorobis 4-fluorophenyl methane, 4,4'-difluorodiphenylmethylchloride, chloro bis-4-fluorophenyl methane, bis 4-fluorophenyl chloromethane, 4,4'-difluorobenzhydrylchloride, 1-chloro 4-fluorophenyl methyl-4-fluorobenzene, 1-chloro-4-fluorophenyl methyl-4-fluorobenzene
PubChem CID 2723773
IUPAC Name 1-[chloro-(4-fluorophenyl)methyl]-4-fluorobenzene
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)F)Cl)F

Specifications

Density 1.28
Boiling Point 126°C to 128°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.557
Quantity 5 g
UN Number UN1760
Beilstein 2052680
Sensitivity Moisture sensitive
Formula Weight 238.66
Percent Purity 98%
Chemical Name or Material 4,4'-Difluorobenzhydryl chloride
Show More Show Less
Hazard Category H227-H302-H315-H319
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 248-201-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only