missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorobenzophenone 99.0+%, TCI America™

Catalog No. p-7126829
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D162125G 25 g
D1621100G 100 g
D1621500G 500 g
3 options

Catalog No. D162125G

Supplier: TCI America D162125G

Only null left

Chemical Identifiers

CAS 90-98-2
Molecular Formula C13H8Cl2O
Molecular Weight (g/mol) 251.11
MDL Number MFCD00000623
InChI Key OKISUZLXOYGIFP-UHFFFAOYSA-N
Synonym 4,4'-dichlorobenzophenone, bis 4-chlorophenyl methanone, methanone, bis 4-chlorophenyl, bis 4-chlorophenyl ketone, dcbp, di p-chlorophenyl ketone, di 4-chlorophenyl ketone, bis p-chlorophenyl ketone, p,p'-dichlorobenzophenone, usaf do-4
PubChem CID 7034
ChEBI CHEBI:27519
IUPAC Name bis(4-chlorophenyl)methanone
SMILES ClC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 147°C
Color White-Yellow
Boiling Point 353°C
Quantity 25 g
Formula Weight 251.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichlorobenzophenone
EINECSNumber (4)-0128
RTECSNumber DJ0525000
TSCA Yes