missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichloro-trans-stilbene 98.0+%, TCI America™

Catalog No. D3063250MG
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3063250MG 250 mg
D30631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3063250MG Supplier TCI America Supplier No. D3063250MG

Chemical Identifiers

CAS 1657-56-3
Molecular Formula C14H10Cl2
Molecular Weight (g/mol) 249.134
InChI Key WELCIURRBCOJDX-OWOJBTEDSA-N
PubChem CID 5352400
IUPAC Name 1-chloro-4-[(E)-2-(4-chlorophenyl)ethenyl]benzene
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)Cl)Cl

Specifications

Melting Point 176°C
Boiling Point 230°C
Quantity 250 mg
Formula Weight 249.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dichloro-trans-stilbene
TSCA No
Recommended Storage Refrigerator