accessibility menu, dialog, popup

UserName

ClCl(E/Z)

CAS RN 1657-56-3

IUPAC Name: 1-chloro-4-[2-(4-chlorophenyl)ethenyl]benzene
Synonyms: 1-chloro-4-[2-(4-chlorophenyl)ethenyl]benzene
Molecular Weight (g/mol): 249.13
Molecular Formula: C14H10Cl2
InChi Key: WELCIURRBCOJDX-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=CC2=CC=C(Cl)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results