missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminooctafluorobiphenyl 97.0+%, TCI America™

Catalog No. d16321g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16321G 1 g
D16325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16321G Supplier TCI America Supplier No. D16321G
Only null left

Chemical Identifiers

CAS 1038-66-0
Molecular Formula C12H4F8N2
Molecular Weight (g/mol) 328.165
MDL Number MFCD00007646
InChI Key FWOLORXQTIGHFX-UHFFFAOYSA-N
Synonym 4,4'-diaminooctafluorobiphenyl, octafluorobenzidine, 4,4'-diaminooctafluorodiphenyl, 2,2',3,3',5,5',6,6'-octafluorobenzidine, 1,1'-biphenyl-4,4'-diamine, 2,2',3,3',5,5',6,6'-octafluoro, 4,4'-diaminoctafluorobiphenyl, benzidine, octafluoro, 4,4'-biphenyldiamine, 2,2',3,3',5,5',6,6'-octafluoro, benzidine, 2,2',3,3',5,5',6,6'-octafluoro, 2,2',3,3',5,5',6,6'-octafluoro-4,4'-bibenzenamine
PubChem CID 70582
IUPAC Name 4-(4-amino-2,3,5,6-tetrafluorophenyl)-2,3,5,6-tetrafluoroaniline
SMILES C1(=C(C(=C(C(=C1F)F)N)F)F)C2=C(C(=C(C(=C2F)F)N)F)F

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 1 g
Formula Weight 328.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diaminooctafluorobiphenyl