accessibility menu, dialog, popup

UserName

CAS RN 1038-66-0

IUPAC Name: octafluoro-[1,1'-biphenyl]-4,4'-diamine
Synonyms: octafluoro-[1,1'-biphenyl]-4,4'-diamine
Molecular Weight (g/mol): 328.17
Molecular Formula: C12H4F8N2
InChi Key: FWOLORXQTIGHFX-UHFFFAOYSA-N
SMILES: NC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(N)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results