missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminodiphenylmethane 98.0+%, TCI America™

Catalog No. M022025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M022025G 25 g
M0220500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M022025G Supplier TCI America Supplier No. M022025G

Chemical Identifiers

CAS 101-77-9
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.27
MDL Number MFCD00007914
InChI Key YBRVSVVVWCFQMG-UHFFFAOYSA-N
Synonym 4,4'-methylenedianiline, 4,4'-diaminodiphenylmethane, dadpm, methylenedianiline, dianilinomethane, tonox, p,p'-methylenedianiline, dianilinemethane, bis 4-aminophenyl methane, 4-4-aminobenzyl aniline
PubChem CID 7577
ChEBI CHEBI:32506
IUPAC Name 4-[(4-aminophenyl)methyl]aniline
SMILES NC1=CC=C(CC2=CC=C(N)C=C2)C=C1

Specifications

Melting Point 91°C
Color White-Yellow
Boiling Point 257°C
Quantity 25 g
UN Number 2651
Formula Weight 198.27
Percent Purity ≥98.0% (T)
Physical Form Pellets
Chemical Name or Material 4,4′-Diaminodiphenylmethane
EINECSNumber (4)-0040
RTECSNumber BY5425000
TSCA Yes