accessibility menu, dialog, popup

UserName

NH2H2N

CAS RN 101-77-9

IUPAC Name: 4-[(4-aminophenyl)methyl]aniline
Synonyms: 4,4'-diaminodiphenylmethane 4,4'-diaminodiphenylmethan jeffamine AP-20 sumicure M epikure ddm MDA α-(p-aminophenyl)-p-toluidine bis-p-aminofenylmethan dianilinomethane p,p'-diaminodiphenylmethane
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: YBRVSVVVWCFQMG-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)