missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminodiphenyl sulfone, 98%

Catalog No. p-7024395
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1897622 100 g
AAA1897636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1897622 Supplier Thermo Scientific Chemicals Supplier No. A1897622
Only null left

CAS: 80-08-0 | C12H12N2O2S | 248.3 g/mol

It is active against a wide range of bacteria, but is mainly employed for its actions against Mycobacterium leprae, being used as part of multidrug regimens in the treatment of all forms of leprosy. It is a second-line medication for the treatment and prevention of Pneumocystis pneumonia and for the prevention of toxoplasmosis in those who have poor immune function. Additionally, it has been used for acne as well as other skin conditions. Moreover it is employed as an anti-inflammatory drug, anti-infective drug, a leprostatic drug and an antimalarial drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water. Solubility in hot methanol is almost transparent. Soluble in DMSO, ethanol and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 80-08-0
Molecular Formula C12H12N2O2S
Molecular Weight (g/mol) 248.3
MDL Number MFCD00007887
InChI Key MQJKPEGWNLWLTK-UHFFFAOYSA-N
Synonym dapsone, diaphenylsulfone, 4,4'-diaminodiphenyl sulfone, dapson, 4,4'-sulfonyldianiline, diaphenylsulfon, sulfona, 4-aminophenyl sulfone, avlosulfone, diphenasone
PubChem CID 2955
ChEBI CHEBI:4325
IUPAC Name 4-(4-aminophenyl)sulfonylaniline
SMILES C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 175°C to 178°C
Odor Odorless
Quantity 100 g
Beilstein 788055
Merck Index 14,2822
Solubility Information Insoluble in water. Solubility in hot methanol is almost transparent. Soluble in DMSO,ethanol and acetone.
Formula Weight 248.3
Percent Purity 98%
Chemical Name or Material 4,4'-Diaminodiphenyl sulfone
Hazard Category H302-H373
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P301+P312-P314-P330-P501c
EINECSNumber 201-248-4
RTECSNumber BY8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only