missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Diaminobenzophenone 98.0+%, TCI America™

Catalog No. D15611G
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D15611G 1 g
D15615G 5 g
2 options

Catalog No. D15611G

Supplier: TCI America D15611G

Only null left

Chemical Identifiers

CAS 611-98-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.252
MDL Number MFCD00038138
InChI Key ZLSMCQSGRWNEGX-UHFFFAOYSA-N
PubChem CID 69149
IUPAC Name bis(4-aminophenyl)methanone
SMILES C1=CC(=CC=C1C(=O)C2=CC=C(C=C2)N)N

Specifications

Melting Point 245°C
Quantity 1 g
Formula Weight 212.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Diaminobenzophenone
EINECSNumber (4)-0154
TSCA No