accessibility menu, dialog, popup

UserName

CAS RN 611-98-3

NH2OH2N

CAS RN 611-98-3

IUPAC Name: 4-(4-aminobenzoyl)aniline
Synonyms: 4-(4-aminobenzoyl)aniline
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: ZLSMCQSGRWNEGX-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results