missing translation for 'onlineSavingsMsg'
Learn More

4,4″-Diamino-p-terphenyl 98.0+%, TCI America™

Catalog No. p-7127711
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33901G 1 g
D33905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33901G Supplier TCI America Supplier No. D33901G
Only null left

Chemical Identifiers

CAS 3365-85-3
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00051532
InChI Key QBSMHWVGUPQNJJ-UHFFFAOYSA-N
Synonym 4,4-diamino-p-terphenyl, 4,4-diaminoterphenyl, 4-4-4-aminophenyl phenyl aniline, ccris 4965, p-terphenyl, 4,4-diamine, 1,1':4',1-terphenyl-4,4-diamine, 4'-4-aminophenyl-1,1'-biphenyl-4-amine, 4,4-diamino-4-terphenyl
PubChem CID 104949
IUPAC Name 4-[4-(4-aminophenyl)phenyl]aniline
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)N)C3=CC=C(C=C3)N

Specifications

Melting Point 242°C
Quantity 1 g
Formula Weight 260.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′′-Diamino-p-terphenyl