accessibility menu, dialog, popup

UserName

CAS RN 3365-85-3

IUPAC Name: 4'-(4-aminophenyl)-[1,1'-biphenyl]-4-amine
Synonyms: 4'-(4-aminophenyl)-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 260.34
Molecular Formula: C18H16N2
InChi Key: QBSMHWVGUPQNJJ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)