missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Di(4-pyridyl)biphenyl 95.0+%, TCI America™
$236.73 - $823.90
Chemical Identifiers
| CAS | 319430-87-0 |
|---|---|
| Molecular Formula | C22H16N2 |
| Molecular Weight (g/mol) | 308.38 |
| MDL Number | MFCD11977635 |
| InChI Key | RERPRBPQDPHWCZ-UHFFFAOYSA-N |
| Synonym | 4,4′C-Bis(4-pyridyl)biphenyl |
| PubChem CID | 15522155 |
| IUPAC Name | 4-[4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl]pyridine |
| SMILES | C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4203200MG
|
TCI America
D4203200MG |
200 mg |
Each for $236.73
|
|
|||||
|
D42031G
|
TCI America
D42031G |
1 g |
Each for $823.90
|
|
|||||
Chemical Identifiers
| 319430-87-0 | |
| 308.38 | |
| RERPRBPQDPHWCZ-UHFFFAOYSA-N | |
| 15522155 | |
| C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=NC=C1 |
| C22H16N2 | |
| MFCD11977635 | |
| 4,4′C-Bis(4-pyridyl)biphenyl | |
| 4-[4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl]pyridine |
Specifications
| 319430-87-0 | |
| Yellow | |
| MFCD11977635 | |
| 4,4′C-Bis(4-pyridyl)biphenyl | |
| C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=NC=C1 | |
| 308.38 | |
| 308.38 | |
| Crystalline Powder |
| 308°C | |
| C22H16N2 | |
| 200 mg | |
| RERPRBPQDPHWCZ-UHFFFAOYSA-N | |
| 4-[4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl]pyridine | |
| 15522155 | |
| ≥95.0% (GC) | |
| 4,4′-Di(4-pyridyl)biphenyl |
Safety and Handling
TSCA : No