accessibility menu, dialog, popup

UserName

CAS RN 319430-87-0

NN

CAS RN 319430-87-0

IUPAC Name: 4-[4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl]pyridine
Synonyms: 4-[4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl]pyridine
Molecular Weight (g/mol): 308.38
Molecular Formula: C22H16N2
InChi Key: RERPRBPQDPHWCZ-UHFFFAOYSA-N
SMILES: C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results