missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Di(4-pyridyl)biphenyl 95.0+%, TCI America™
Supplier: TCI America D42031G
Specifications
| 4,4′-Di(4-pyridyl)biphenyl | |
| 308°C | |
| C22H16N2 | |
| 1 g | |
| RERPRBPQDPHWCZ-UHFFFAOYSA-N | |
| 4-[4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl]pyridine | |
| 15522155 | |
| ≥95.0% (GC) |
| 319430-87-0 | |
| Yellow | |
| MFCD11977635 | |
| 4,4′C-Bis(4-pyridyl)biphenyl | |
| C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=NC=C1 | |
| 308.38 | |
| 308.38 | |
| Crystalline Powder |
Chemical Identifiers
| 319430-87-0 | |
| 308.38 | |
| RERPRBPQDPHWCZ-UHFFFAOYSA-N | |
| 15522155 | |
| C1=CC(=CC=N1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=NC=C1 |
| C22H16N2 | |
| MFCD11977635 | |
| 4,4′C-Bis(4-pyridyl)biphenyl | |
| 4-[4'-(pyridin-4-yl)-[1,1'-biphenyl]-4-yl]pyridine |
Safety and Handling
TSCA : No