missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Butylidenebis(6-tert-butyl-m-cresol) 97.0+%, TCI America™

Catalog No. p-7118639
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B072425G 25 g
B0724500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B072425G Supplier TCI America Supplier No. B072425G

Chemical Identifiers

CAS 85-60-9
Molecular Formula C26H38O2
Molecular Weight (g/mol) 382.59
MDL Number MFCD00026289
InChI Key PFANXOISJYKQRP-UHFFFAOYSA-N
Synonym santowhite, santowhite powder, sumilit bbm, sumilizer bbm, anullex pba 15, 4,4'-butylidenebis 6-tert-butyl-m-cresol, yoshinox bb, sumilizer bbm-s, swp antioxidant, noclizer ns 30
PubChem CID 6815
IUPAC Name 2-tert-butyl-4-[1-(5-tert-butyl-4-hydroxy-2-methylphenyl)butyl]-5-methylphenol
SMILES CCCC(C1=CC(=C(O)C=C1C)C(C)(C)C)C1=CC(=C(O)C=C1C)C(C)(C)C

Specifications

Melting Point 211°C
Color White
Quantity 25 g
Formula Weight 382.59
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Butylidenebis(6-tert-butyl-m-cresol)
EINECSNumber (4)-0250
RTECSNumber GO7050000
TSCA Yes