missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Bromophenyl)butyric Acid 98.0+%, TCI America™

Catalog No. p-7119973
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31311G 1 g
B31315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31311G Supplier TCI America Supplier No. B31311G

Chemical Identifiers

CAS 35656-89-4
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD00463170
InChI Key AGIIMNQWNPUJPT-UHFFFAOYSA-N
PubChem CID 252732
IUPAC Name 4-(4-bromophenyl)butanoic acid
SMILES C1=CC(=CC=C1CCCC(=O)O)Br

Specifications

Melting Point 72°C
Boiling Point 176°C
Quantity 1 g
Synonym 4-4-bromophenyl butanoic acid, 4-4-bromophenyl butyric acid, benzenebutanoic acid, 4-bromo, 4-bromo-benzenebutanoic acid, pubchem16174, acmc-209iig, 4-bromobenzenebutanoic acid, benzenebutanoic acid,4-bromo, 4-4-bromo-phenyl-butyric acid
Formula Weight 243.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Bromophenyl)butyric Acid