accessibility menu, dialog, popup

UserName

HOOBr

CAS RN 35656-89-4

IUPAC Name: 4-(4-bromophenyl)butanoic acid
Synonyms: 4-(4-bromophenyl)butanoic acid
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: AGIIMNQWNPUJPT-UHFFFAOYSA-N
SMILES: OC(=O)CCCC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results