missing translation for 'onlineSavingsMsg'
Learn More

4'-(4-Bromophenyl)acetophenone, 97%

Catalog No. AAL1379814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1379814 25 g
AAL1379822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1379814 Supplier Thermo Scientific Chemicals Supplier No. L1379814

CAS: 5731-01-1 | C14H11BrO | 275.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5731-01-1
MDL Number MFCD00143242

Specifications

Melting Point 125°C to 127°C
Molecular Formula C14H11BrO
Quantity 25 g
Beilstein 2048590
Synonym 4-acetyl-4'-bromobiphenyl, 4'-4-bromophenyl acetophenone, 1-4-4-bromophenyl phenyl ethanone, 4'-p-bromophenyl acetophenone, 1-4'-bromo-1,1'-biphenyl-4-yl ethanone, 4-acetyl-4'-bromodiphenyl, 4-bromo-4'-acetylbiphenyl, ethanone, 1-4'-bromo 1,1'-biphenyl-4-yl, 1-4'-bromo 1,1'-biphenyl-4-yl ethanone, 1-4'-bromo 1,1'-biphenyl-4-yl ethan-1-one
InChI Key UUVKNCRMWPNBNM-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)Br
IUPAC Name 1-[4-(4-bromophenyl)phenyl]ethanone
Molecular Weight (g/mol) 275.15
PubChem CID 79806
Formula Weight 275.15
Percent Purity 97%
Chemical Name or Material 4'-(4-Bromophenyl)acetophenone
Show More Show Less
EINECSNumber 227-236-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only