accessibility menu, dialog, popup

UserName

CAS RN 5731-01-1

IUPAC Name: 1-{4'-bromo-[1,1'-biphenyl]-4-yl}ethan-1-one
Synonyms: 1-{4'-bromo-[1,1'-biphenyl]-4-yl}ethanone
Molecular Weight (g/mol): 275.15
Molecular Formula: C14H11BrO
InChi Key: UUVKNCRMWPNBNM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)