missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(chloromethyl)biphenyl 95.0+%, TCI America™

Catalog No. p-7119477
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B209925G 25 g
B2099500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B209925G Supplier TCI America Supplier No. B209925G
Only null left

Chemical Identifiers

CAS 1667-10-3
Molecular Formula C14H12Cl2
Molecular Weight (g/mol) 251.15
MDL Number MFCD00674019
InChI Key INZDTEICWPZYJM-UHFFFAOYSA-N
PubChem CID 74275
IUPAC Name 4,4'-bis(chloromethyl)-1,1'-biphenyl
SMILES ClCC1=CC=C(C=C1)C1=CC=C(CCl)C=C1

Specifications

Melting Point 141°C
Color White-Yellow
Boiling Point 184°C
Quantity 25 g
UN Number 3261
Formula Weight 251.15
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(chloromethyl)biphenyl
EINECSNumber (4)-0798
TSCA No